Learn More
Sino Biological Recombinant Mouse GRO gamma/CXCL3 Protein (His Tag)
Supplier:  Sino Biological 50258M08Y1
Encompass_Preferred
A DNA sequence encoding the mouse Cxcl3 (NP_976065.1) (Ala28-Ser100) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3324