Learn More
Kingfisher Biotech Inc Mouse CXCL7 Recombinant Protein (Yeast, Endotoxin - Free) - 500 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1935M500
Encompass
The Mouse CXCL7 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Mouse CXCL7 applications are for cell culture, ELISA standard, and Western Blot Control. Mouse CXCL7 yeast-derived recombinant protein can be purchased in multiple sizes. Mouse CXCL7 Specifications: (Molecular Weight: 8.1 kDa) (Amino Acid Sequence: KSDGMDPYIE LRCRCTNTIS GIPFNSISLV NVYRPGVHCA DVEVIATLKN GQKTCLDPNA PGVKRIVMKI LEGY (74)) (Gene ID: 57349). For research use only. Made in the USA
Catalog No. 50-253-2586