Learn More
Abcam Mouse CXCL9 ELISA Kit, 1X96TEST
Supplier:  Abcam AB2033641X96TESTS
Encompass_Preferred
The Abcam Mouse CXCL9 ELISA Kit is a rapid and sensitive SimpleStep Sandwich ELISA designed for quantitative measurement of Mouse CXCL9 protein. This kit simplifies the experimental procedure with a single-step incubation method, significantly reducing assay time. With a sensitivity of 1.4 pg/mL, it is ideal for detecting low concentrations of Mouse CXCL9 in various biological samples and compatible with a range of sample types, making it versatile for research use.
  • Delivers results in just 90 minutes
  • Achieves a sensitivity of 1.4 pg/mL
  • Validated for various sample types: serum, plasma, cell culture supernatant, and tissue extracts
  • Employs straightforward mix-wash-read protocol
  • Format: 12 x 8 well strips
  • Range: 6.25 - 400 pg/mL
  • For research use only
Catalog No. NC2255910