Learn More
Sino Biological Anti-CXCL9/MIG Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50155T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CXCL9 / MIG (rM CXCL9 / MIG; Catalog#50155-MNAE; P18340; Thr22-Thr126). CXCL9 / MIG specific IgG was purified by Mouse CXCL9 / MIG affinity chromatography.
Catalog No. 50-241-2483