Learn More
BIOHIPPO Mouse CXCR6 Microsample
SDP
Catalog No. 5000314329
Encompass
Mouse CXCR6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CXCR6 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 061 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CXCR6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CXCR6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CXCR6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14329 Supplier BIOHIPPO Supplier No. BHE15212447
Mouse CXCR6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CXCR6 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 061 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CXCR6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CXCR6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CXCR6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color