Learn More
Biomatik Corporation Mouse Cyclophilin A (CYPA) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10920
Encompass_Preferred
Mouse Cyclophilin A (CYPA) High-sensitive ELISA Kit for detecting CYPA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.64ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17742. Gene ID: 268373. Target Synonyms: PPIA; CyP-A; CYPH; Peptidylprolyl Isomerase A; Peptidyl-Prolyl Isomerase A; Peptidyl-prolyl cis-trans isomerase A; Cyclosporin A-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8267