Learn More
Biomatik Corporation Mouse Cystatin C, Cys-C Elisa Kit
Supplier:  Biomatik Corporation EKC31389
Encompass_Preferred
Mouse Cystatin C, Cys-C Elisa Kit for detecting Cystatin C in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.5-160ng/ml. Sensitivity: 0.819ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P21460; Cst3. Target Synonyms: Cst3Cystatin-C; Cystatin-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7286