Learn More
Biomatik Corporation Mouse Cytochrome b-245 Beta Polypeptide (CYBb) ELISA Kit
Supplier:  Biomatik Corporation EKU12432
Encompass_Preferred
Mouse Cytochrome b-245 Beta Polypeptide (CYBb) ELISA Kit for detecting CYBb in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61093. Gene ID: 13058. Target Synonyms: CGD; GP91PHOX; NOX2; Neutrophil cytochrome b91; Cytochrome b558 beta; Heme-binding membrane glycoprotein gp91phox; Superoxide-generating NADPH oxidase heavy chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6677