Learn More
Biomatik Corporation Mouse Cytochrome P450 26B1 (CYP26B1) ELISA Kit
Supplier:  Biomatik Corporation EKU12434
Encompass_Preferred
Mouse Cytochrome P450 26B1 (CYP26B1) ELISA Kit for detecting CYP26B1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q811W2. Gene ID: 232174. Target Synonyms: CYP26A2; P450RAI-2; Cytochrome P450 Family 26 Subfamily B Polypeptide 1; Cytochrome P450 retinoic acid-inactivating 2; Retinoic acid-metabolizing cytochrome. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7966