Learn More
American Research Products Inc Mouse Cytochrome P450 2E1 CYP
Catalog No. 5000328008
Encompass
Mouse Cytochrome P450 2E1 (CYP2E1) ELISA kit from CUSABIO Detection Range 39 pg/mL-2500 pg/mL Sensitivity 9 75 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28008 Supplier American Research Products Inc Supplier No. CSBEL006425MO
Mouse Cytochrome P450 2E1 (CYP2E1) ELISA kit from CUSABIO Detection Range 39 pg/mL-2500 pg/mL Sensitivity 9 75 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich