Learn More
Raybiotech Inc MOUSE CYTOKINE ARRAY C1 (8)
Catalog No. 501043199
Encompass_Preferred
RayBio C-Series Mouse Cytokine Antibody Array 1 Kit. Detects 22 Mouse Cytokines. Suitable for all liquid sample types.
Catalog No. 50-104-3199 Supplier Raybiotech Inc Supplier No. AAMCYT18
RayBio C-Series Mouse Cytokine Antibody Array 1 Kit. Detects 22 Mouse Cytokines. Suitable for all liquid sample types.