Learn More
Raybiotech Inc C-Series Mouse Cytokine Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAMCYT12
Encompass_Preferred
Detects 22 Mouse Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: GCSF, GM-CSF, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17A, IFN-gamma, MCP-1 (CCL2), MCP-5, RANTES (CCL5), SCF, TNF RI (TNFRSF1A), TNF alpha, Thrombopoietin (TPO), VEGF-A
Catalog No. 50-194-8696