Learn More
Raybiotech Inc C-Series Mouse Cytokine Antibody Array 2 Kit.
Catalog No. 501948702
Encompass_Preferred
Detects 32 Mouse Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: 6Ckine (CCL21), CTACK (CCL27), Eotaxin-1 (CCL11), GCSF, GM-CSF, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17A, IFN-gamma, KC (CXCL1), Leptin, MCP-1 (CCL2), MCP-5, MIP-1 alpha (CCL3), MIP-2, MIP-3 beta (CCL19), RANTES (CCL5), SCF, TNF RI (TNFRSF1A), TARC (CCL17), TIMP-1, TNF alpha, Thrombopoietin (TPO), VEGF-A
Catalog No. 50-194-8702 Supplier Raybiotech Inc Supplier No. AAMCYT22
Detects 32 Mouse Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: 6Ckine (CCL21), CTACK (CCL27), Eotaxin-1 (CCL11), GCSF, GM-CSF, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17A, IFN-gamma, KC (CXCL1), Leptin, MCP-1 (CCL2), MCP-5, MIP-1 alpha (CCL3), MIP-2, MIP-3 beta (CCL19), RANTES (CCL5), SCF, TNF RI (TNFRSF1A), TARC (CCL17), TIMP-1, TNF alpha, Thrombopoietin (TPO), VEGF-A