Learn More
Raybiotech Inc MOUSE CYTOKINE ARRAY C2000 (4)
Supplier:  Raybiotech Inc AAMCYT20004
Encompass_Preferred
RayBio C-Series Mouse Cytokine Antibody Array 2000. A combination of RayBio Mouse Cytokine Antibody Array C3, C4, & C5 Kit. Detects 144 Mouse Cytokines. Suitable for all liquid sample types.
Catalog No. 50-104-3201