Learn More
Raybiotech Inc MOUSE CYTOKINE ARRAY C3 (4)
Catalog No. 501043207
Encompass_Preferred
RayBio C-Series Mouse Cytokine Antibody Array 3 Kit. Detects 62 Mouse Cytokines. Suitable for all liquid sample types.
Catalog No. 50-104-3207 Supplier Raybiotech Inc Supplier No. AAMCYT34
RayBio C-Series Mouse Cytokine Antibody Array 3 Kit. Detects 62 Mouse Cytokines. Suitable for all liquid sample types.