Learn More
Raybiotech Inc C-Series Mouse Cytokine Antibody Array 3 Kit.
Catalog No. 501948705
Encompass_Preferred
Detects 62 Mouse Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. View protein list on RayBiotech’s website – AAM-CYT-3-2
Catalog No. 50-194-8705 Supplier Raybiotech Inc Supplier No. AAMCYT32
Detects 62 Mouse Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. View protein list on RayBiotech’s website – AAM-CYT-3-2