Learn More
Raybiotech Inc C-Series Mouse Cytokine Antibody Array 4 Kit.
Supplier:  Raybiotech Inc AAMCYT48
Encompass_Preferred
Detects 34 Mouse Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: bFGF, CD26 (DPPIV), Dtk, E-Selectin, Fc gamma RIIB (CD32b), Flt-3 Ligand, GITR (TNFRSF18), HGFR, ICAM-1 (CD54), IGFBP-2, IGF-1, IGF-2, IL-15, IL-17 RB, IL-7, I-TAC (CXCL11), Lungkine (CXCL15), MDC (CCL22), MMP-2, MMP-3, Osteopontin (SPP1), Osteoprotegerin (TNFRSF11B), Pro-MMP-9, Resistin, Sonic Hedgehog N-Terminal (Shh-N), TCK-1 (CXCL7), TIMP-2, TRANCE (TNFSF11), TROY (TNFRSF19), TSLP, VEGFR1, VEGFR2, VEGFR3, VEGF-D
Catalog No. 50-194-8710