Learn More
Raybiotech Inc C-Series Mouse Cytokine Antibody Array 5 Kit.
Supplier:  Raybiotech Inc AAMCYT54
Encompass_Preferred
Detects 48 Mouse Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: 4-1BB (TNFRSF9/CD137), 6Ckine (CCL21), ACE, ALK-1, Amphiregulin, Cardiotrophin-1 (CT-1), CD27 (TNFRSF7), CD27 Ligand (TNFSF7), CD36 (SR-B3), CD40 Ligand (TNFSF5), Chordin, CTLA-4 (CD152), Decorin, DKK-1, E-Cadherin, EGF, Endoglin (CD105), Epigen, Epiregulin, Galectin-1, Gas 1, Gas 6, GITR Ligand (TNFSF18), Granzyme B, HAI-1, HGF, IL-1 R4 (ST2), IL-11, IL-17B, IL-17E (IL-25), IL-17F, IL-1 RA (IL-1 F3), IL-2 R alpha, IL-20, IL-21, IL-28A (IFN-lambda 2), IL-6 R, JAM-A (CD321/F11R), MAdCAM-1, MFG-E8, Neprilysin, Pentraxin-3 (TSG-14), Prolactin, RAGE, TACI (TNFRSF13B), TREM-1, TWEAK (TNFSF12), TWEAK R (TNFRSF12)
Catalog No. 50-194-8712