Learn More
Raybiotech Inc C-Series Mouse Cytokine Antibody Array 6 Kit.
Supplier:  Raybiotech Inc AAMCYT68
Encompass_Preferred
Detects 97 Mouse Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. View protein list on RayBiotech’s website – AAM-CYT-6-8
Catalog No. 50-194-8716