Learn More
Raybiotech Inc C-Series Mouse Cytokine Antibody Array 7 Kit.
Supplier:  Raybiotech Inc AAMCYT74
Encompass_Preferred
Detects 40 Mouse Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: CD80 (B7-1), BAFF R, Betacellulin (BTC), C5a, CCL6, CD48 (SLAMF2), CD6, Chemerin, Clusterin, Lungkine (CXCL15), Cystatin C, DAN, DLL4, EDAR, Endocan, Fetuin A, H60, IL-33 (IL-1 F11), IL-7 R alpha, Kremen-1, Limitin, Lipocalin-2 (NGAL), LOX-1, Marapsin, MBL-2, Meteorin, Nope, NOV (CCN3), Osteoactivin (GPNMB), OX40 Ligand, P-Cadherin, Periostin, PLGF-2, Progranulin, Prostasin, Renin 1, Testican 3 (SPOCK3), TIM-1 (KIM-1), TRAIL (TNFSF10), Tryptase epsilon
Catalog No. 50-194-8718