Learn More
Raybiotech Inc G-Series Mouse Cytokine Antibody Array 1000.
Supplier:  Raybiotech Inc AAMCYTG10008
Encompass_Preferred
A combination of Mouse Cytokine Antibody Array G3 & G4. Detects 96 Mouse Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – AAM-CYT-G1000-8
Catalog No. 50-194-8721