Learn More
Raybiotech Inc G-Series Mouse Cytokine Antibody Array 3 Kit.
Catalog No. 501948726
Encompass_Preferred
Detects 62 Mouse Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – AAM-CYT-G3-4
Catalog No. 50-194-8726 Supplier Raybiotech Inc Supplier No. AAMCYTG34
Detects 62 Mouse Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – AAM-CYT-G3-4