Learn More
Raybiotech Inc G-Series Mouse Cytokine Array 1 Kit.
Catalog No. 501950450
Encompass_Preferred
Semi-quantitative measurement of 20 mouse cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: GM-CSF, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-10, IL-12 p70, IL-13, IL-17A, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, KC (CXCL1), MCP-1 (CCL2), M-CSF, RANTES (CCL5), TNF alpha, VEGF-A
Catalog No. 50-195-0450 Supplier Raybiotech Inc Supplier No. GSMCYT11
Semi-quantitative measurement of 20 mouse cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: GM-CSF, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-10, IL-12 p70, IL-13, IL-17A, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, KC (CXCL1), MCP-1 (CCL2), M-CSF, RANTES (CCL5), TNF alpha, VEGF-A