Learn More
Raybiotech Inc G-Series Mouse Cytokine Array 4 Kit.
Supplier:  Raybiotech Inc GSMCYT41
Encompass_Preferred
Semi-quantitative measurement of 40 mouse cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Amphiregulin, Axl, CD27 Ligand (TNFSF7), CD30 (TNFRSF8), CD40 (TNFRSF5), CXCL16, EGF, E-Selectin, Fractalkine (CX3CL1), GITR (TNFRSF18), HGF, IGF-1, IGFBP-2, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 ra (IL-1 F3), IL-12 p70, IL-17E (IL-25), IL-17F, IL-2 R alpha, IL-20, IL-23 p19, IL-28A (IFN-lambda 2), I-TAC (CXCL11), MDC (CCL22), MIP-2, MIP-3 alpha (CCL20), Osteopontin (SPP1), Osteoprotegerin (TNFRSF11B), Prolactin, Pro-MMP-9, P-Selectin, Resistin, SCF, SDF-1 alpha (CXCL12 alpha), Thrombopoietin (TPO), VCAM-1 (CD106), VEGF-A, VEGF-D
Catalog No. 50-195-0452