Learn More
Raybiotech Inc G-Series Mouse Cytokine Array 4000 Kit.
Supplier:  Raybiotech Inc GSMCAA40001
Encompass_Preferred
A combination of 5 non-overlapping arrays to measure the relative expression levels of 200 mouse cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – GSM-CAA-4000-1
Catalog No. 50-195-0446