Learn More
Raybiotech Inc G-Series Mouse Cytokine Array 7 Kit.
Supplier:  Raybiotech Inc GSMCYT72
Encompass_Preferred
Semi-quantitative measurement of 40 mouse cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: BAFF R, Betacellulin (BTC), C5a, CCL6, CD48 (SLAMF2), CD6, CD80 (B7-1), Chemerin, Clusterin, Cystatin C, DAN, DLL4, EDAR, Endocan, Fetuin A, H60, IL-33 (IL-1 F11), IL-7 R alpha, Kremen-1, Limitin, Lipocalin-2 (NGAL), LOX-1, Lungkine (CXCL15), Marapsin, MBL-2, Meteorin, Nope, NOV (CCN3), Osteoactivin (GPNMB), OX40 Ligand, P-Cadherin, Periostin, PlGF-2, Progranulin, Prostasin, Renin 1, Testican 3, TIM-1 (KIM-1), TRAIL (TNFSF10), Tryptase epsilon
Catalog No. 50-195-0459