Learn More
Raybiotech Inc G-Series Mouse Cytokine Array 8 Kit.
Supplier: Raybiotech Inc GSMCYT82
Semi-quantitative measurement of 40 Mouse cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: 6Ckine (CCL21), Activin A, ADAMTS-1 (METH1), Adiponectin (ACRP30), Angiopoietin-3 (ANGPTL1), ANGPTL3, Artemin, CCL28 (MEC), CD36 (SR-B3), Chordin, CRP (C-Reactive Protein), E-Cadherin, Epigen, Epiregulin, Fas (TNFRSF6/Apo-1), Galectin-7, gp130, Granzyme B, Gremlin-1, IFN-gamma R1, IL-17 RB, IL-17B, IL-22, MIP-1 beta (CCL4), MMP-10, MMP-2, MMP-3, PDGF-AA, Persephin, sFRP-3, SLAM (CD150), Sonic Hedgehog N-Terminal (Shh-N), TCK-1 (CXCL7), TECK (CCL25), TGF beta 1, Trance (TNFSF11), TREML1 (TLT-1), TWEAK (TNFSF12), VEGF-B, VEGFR2