Learn More
Raybiotech Inc Mouse Cytokine Array Kit, 20 cytokines detected, 50 μL sample volume, 6 hours assay time, glass slide format, high-throughput
Catalog No. NC3641063
Encompass_Preferred
Mouse Cytokine Array Kit, 20 cytokines detected, 50 μL sample volume, 6 hours assay time, glass slide format, high-throughput
Catalog No. NC3641063 Supplier Raybiotech Inc Supplier No. QAMCYT14
Mouse Cytokine Array Kit, 20 cytokines detected, 50 μL sample volume, 6 hours assay time, glass slide format, high-throughput