Learn More
Raybiotech Inc Quantibody Mouse Cytokine Array 1 Kit.
Supplier:  Raybiotech Inc QAMCYT11
Encompass_Preferred
Detects 20 Mouse Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: GM-CSF, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-10, IL-12 p70, IL-13, IL-17A, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, KC (CXCL1), MCP-1 (CCL2), M-CSF, RANTES (CCL5), TNF alpha, VEGF-A
Catalog No. 50-195-0871