Learn More
Raybiotech Inc Quantibody Mouse Cytokine Array 1000 Kit.
Catalog No. 501950861
Encompass_Preferred
A combination of Mouse Cytokine Array Q4 and Mouse Cytokine Array Q5. Detects 80 Mouse Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. View protein list on RayBiotech’s website – QAM-CAA-1000-1
Catalog No. 50-195-0861 Supplier Raybiotech Inc Supplier No. QAMCAA10001
A combination of Mouse Cytokine Array Q4 and Mouse Cytokine Array Q5. Detects 80 Mouse Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. View protein list on RayBiotech’s website – QAM-CAA-1000-1