Learn More
Raybiotech Inc Quantibody Mouse Cytokine Array 2000 Kit.
Supplier:  Raybiotech Inc QAMCAA20002
Encompass_Preferred
A combination of Mouse Cytokine Array Q4, Mouse Cytokine Array Q5, and Mouse Cytokine Array Q6. Detects 120 Mouse Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. View protein list on RayBiotech’s website – QAM-CAA-2000-2
Catalog No. 50-195-0864