Learn More
Raybiotech Inc Quantibody Mouse Cytokine Array 3000 Kit.
Supplier:  Raybiotech Inc QAMCAA30001
Encompass_Preferred
A combination of Mouse Cytokine Array Q4, Mouse Cytokine Array Q5, Mouse Cytokine Array Q6, and Mouse Cytokine Array Q7. Detects 160 Mouse Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. View protein list on RayBiotech’s website – QAM-CAA-3000-1
Catalog No. 50-195-0865