Learn More
Raybiotech Inc Quantibody Mouse Cytokine Array 4 Kit.
Supplier:  Raybiotech Inc QAMCYT42
Encompass_Preferred
Detects 40 Mouse Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: Amphiregulin, Axl, CD27 Ligand (TNFSF7), CD30 (TNFRSF8), CD40 (TNFRSF5), CXCL16, EGF, E-Selectin, Fractalkine (CX3CL1), GITR (TNFRSF18), HGF, IGF-1, IGFBP-2, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 ra (IL-1 F3), IL-12 p70, IL-17E (IL-25), IL-17F, IL-2 R alpha, IL-20, IL-23 p19, IL-28A (IFN-lambda 2), I-TAC (CXCL11), MDC (CCL22), MIP-2, MIP-3 alpha (CCL20), Osteopontin (SPP1), Osteoprotegerin (TNFRSF11B), Prolactin, Pro-MMP-9, P-Selectin, Resistin, SCF, SDF-1 alpha (CXCL12 alpha), Thrombopoietin (TPO), VCAM-1 (CD106), VEGF-A, VEGF-D
Catalog No. 50-195-0874