Learn More
Raybiotech Inc Quantibody Mouse Cytokine Array 6 Kit.
Supplier:  Raybiotech Inc QAMCYT62
Encompass_Preferred
Detects 40 Mouse Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: 4-1BB (TNFRSF9/CD137), Ace, ALK-1, Cardiotrophin-1 (CT-1), CD27 (TNFRSF7), CD32a, CD40 Ligand (TNFSF5), CTLA-4 (CD152), Decorin, DKK-1, Dtk, Endoglin (CD105), Flt-3 Ligand, Galectin-1, Galectin-3, Gas 1, Gas 6, GITR Ligand (TNFSF18), HAI-1, HGFR, IL-1 R4 (ST2), IL-3 R beta, IL-9, JAM-A (CD321/F11R), Leptin R, L-Selectin (CD62L), Lymphotactin (XCL1), MAdCAM-1, MFG-E8, MIP-3 beta (CCL19), Neprilysin, Pentraxin-3 (TSG-14), RAGE, TACI (TNFRSF13B), TREM-1, TROY (TNFRSF19), TSLP, TWEAK R (TNFRSF12), VEGFR1, VEGFR3
Catalog No. 50-195-0878