Learn More
Raybiotech Inc Quantibody Mouse Cytokine Array 9 Kit.
Supplier:  Raybiotech Inc QAMCYT91
Encompass_Preferred
Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: 2B4 (CD244), 4-1BB Ligand (TNFSF9), Activin R1B (ALK-4), ADP-Ribosyl Cyclase 2 (BST-1/CD157), Ameloblastin, ANGPTL7, B7-H2 (ICOS Ligand), B7-H3 (CD276), B7-H4, BAFF, C1q R1 (CD93), Cathepsin H, CD117 (c-kit), CD200, CD229 (SLAMF3), CD28, CD300b (LMIR5), CD39 (ENTPD1), CD44, CD45, CD69, CD86 (B7-2), CD99-L2, CHL-1 (L1CAM-2), Chordin-like 2 (CHRDL2), COCO, CRACC (SLAMF7), CXADR, DcTRAIL R1 (TNFRSF23), Dectin-2 (CLEC6A), DNAM-1 (CD226), DNER, Endoglycan (PODXL2), EphA8, EphB2, EphB4, EphB6, Ephrin-A2, Ephrin-A4, FGF-21
Catalog No. 50-195-0883