Learn More
American Research Products Inc Mouse cytotoXic T lymphocyte a
Catalog No. 5000329332
Encompass
Mouse cytotoXic T lymphocyte associated antigen 4 CTLA-4 ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29332 Supplier American Research Products Inc Supplier No. CSBE13637M
Mouse cytotoXic T lymphocyte associated antigen 4 CTLA-4 ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich