Learn More
American Research Products Inc Mouse D Lactate Dehydrogenase
Catalog No. 5000329531
Encompass
Mouse D-Lactate Dehydrogenase D-LDH ELISA Kit from CUSABIO Detection Range 4 69 mU/mL-300 mU/mL Sensitivity 1 17 mU/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29531 Supplier American Research Products Inc Supplier No. CSBE11723M
Mouse D-Lactate Dehydrogenase D-LDH ELISA Kit from CUSABIO Detection Range 4 69 mU/mL-300 mU/mL Sensitivity 1 17 mU/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich