Learn More
Raybiotech Inc Mouse DAN ELISA Kit
Supplier:  Raybiotech Inc ELMDAN1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 35 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects mouse DAN, no cross-reactivity with rhDAN, rhCOCO, rmBMP-4 or rmGremlin
Catalog No. 50-194-9963