Learn More
BIOHIPPO Mouse DAO Microsample
SDP
Catalog No. 5000314735
Encompass
Mouse DAO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of DAO in serum plasma tissue homogenates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 32 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse DAO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse DAO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse DAO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14735 Supplier BIOHIPPO Supplier No. BHE15212529
Mouse DAO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of DAO in serum plasma tissue homogenates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 32 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse DAO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse DAO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse DAO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color