Learn More
Sino Biological Anti-DOPA Decarboxylase/DDC Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50799RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse DOPA Decarboxylase / DDC (Catalog#50799-M08B; O88533; Met1-Glu480). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3080