Learn More
Biomatik Corporation Mouse D-Dimer (D2D) CLIA Kit
Supplier:  Biomatik Corporation EKU09769
Encompass_Preferred
Mouse D-Dimer (D2D) CLIA Kit for detecting D2D in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 195.3-50,000pg/ml. Sensitivity: 76.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Target Synonyms: D 2 Dimer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7349