Learn More
Bon Opus Biosciences Recombinant Mouse Dectin-1 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP109150UG
Encompass_Preferred
Recombinant Mouse Dectin-1 (C-hFc) is produced by Human Cells. The target gene encoding Phe69-Leu244 is expressed with a C-hFc tag. Accession# NP_064392.2(Q6QLQ4). Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1091
Catalog No. 50-217-2689