Learn More
Biomatik Corporation Mouse Defensin Alpha 2 (DEFa2) ELISA Kit
Supplier:  Biomatik Corporation EKU12365
Encompass_Preferred
Mouse Defensin Alpha 2 (DEFa2) ELISA Kit for detecting DEFa2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.54ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28309. Target Synonyms: Defcr2; Defensin-Related Cryptdin-2; Cryptdin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7531