Learn More
Biomatik Corporation Mouse Defensin Alpha 3, Neutrophil Specific (DEFa3) ELISA Kit
Supplier:  Biomatik Corporation EKU12223
Encompass_Preferred
Mouse Defensin Alpha 3, Neutrophil Specific (DEFa3) ELISA Kit for detecting DEFa3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28310. Gene ID: 13237. Target Synonyms: HNP3; DEF3; HNP-2; Neutrophil defensin 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8000