Learn More
Biomatik Corporation Mouse Defensin Alpha 5, Paneth Cell Specific (DEFa5) ELISA Kit
Supplier:  Biomatik Corporation EKU03653
Encompass_Preferred
Mouse Defensin Alpha 5, Paneth Cell Specific (DEFa5) ELISA Kit for detecting DEFa5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28312. Target Synonyms: DEF-A5; DEF5; HD-5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6582