Learn More
Biomatik Corporation Mouse Defensin Beta 2 (DEFb2) ELISA Kit
Supplier:  Biomatik Corporation EKU03670
Encompass_Preferred
Mouse Defensin Beta 2 (DEFb2) ELISA Kit for detecting DEFb2 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P82020. Gene ID: 13215. Target Synonyms: B-DF2; DEFB2; BD2; DEF-B2; HBD2; DEFB102; HBD-2; SAP1; DEFB4A; DEFB4; Skin-Antimicrobial Peptide 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6526