Learn More
Biomatik Corporation Mouse Deleted In Malignant Brain Tumors 1 (DMBT1) ELISA Kit
Supplier:  Biomatik Corporation EKU10788
Encompass_Preferred
Mouse Deleted In Malignant Brain Tumors 1 (DMBT1) ELISA Kit for detecting DMBT1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q60997. Gene ID: 12945. Target Synonyms: GP340; SAG; Muclin; Glycoprotein 340; Hensin; Salivary agglutinin; Surfactant pulmonary-associated D-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6678