Learn More
Sciencell Research Laboratories MOUSE DERMAL FIBROBLASTS C57BL
Catalog No. NC1713317
Encompass
Mouse Dermal Fibroblasts from C57BL/6MDF are isolated from postnatal day 2 C57BL/6 mouse skin. MDF are cryopreserved at passage one and delivered frozen. Each vial contains >5 x 10^5 cells in 1 ml volume.
Catalog No. NC1713317 Supplier Sciencell Research Laboratories Supplier No. M230057
Mouse Dermal Fibroblasts from C57BL/6MDF are isolated from postnatal day 2 C57BL/6 mouse skin. MDF are cryopreserved at passage one and delivered frozen. Each vial contains >5 x 10^5 cells in 1 ml volume.