Learn More
Sino Biological Anti-Desmin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106160T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse Desmin/DES, and purified by antigen affinity chromatography.
Catalog No. 50-241-1067