Learn More
Biocare Medical, LLC Mouse Detective - 25 ml
SDP
Catalog No. 5082923
Encompass_Preferred
Mouse Detective is an immunohistochemical reagent designed for blocking endogenous mouse IgG in mouse tissues and red blood cells. After the application of mouse detective, a mouse monoclonal antibody can be applied followed by a biotinylated mouse link and Streptavidin label used for detection of mouse on mouse. It can also be used on an automated staining system.
Catalog No. 50-829-23 Supplier Biocare Medical, LLC Supplier No. MD975H
Mouse Detective is an immunohistochemical reagent designed for blocking endogenous mouse IgG in mouse tissues and red blood cells. After the application of mouse detective, a mouse monoclonal antibody can be applied followed by a biotinylated mouse link and Streptavidin label used for detection of mouse on mouse. It can also be used on an automated staining system.